Learn More
Eppendorf™ Galaxy™ 48 R Incubators
When a high level of cell culture isolation is required or very limited space is available in the lab, the stackable Galaxy 48R is the right choice. The high temperature disinfection, a seamless chamber and a fanless design makes it easy to clean and provides high contamination protection.
Brand: Eppendorf™ CO48310001
Description
- 48 Liter (1.7 ft3) provides mid-sized capacity within a small footprint
- Options: High Temp. Disinfection (4 hour at 120 °C), O2 control (e.g. for hypoxic studies), 2-split inner glass door
- Fanless design offers more usable space
- Six-sided direct heating system provides a uniform incubated environment
- Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
- Low gas consumption
- Integrated alarm relay
Specifications
UK/HKG Power Supply | |
IR Sensor | |
None | |
Single Glass Inner Door |
Eppendorf Delivery Check list June 2022.pdf
File size: 350.1 kB
Product Content Correction
Product Title
Eppendorf™ Galaxy™ 48 R Incubators > Capacity: 48L