Learn More
Eppendorf™ Galaxy™ 48 R Incubators
Click to view available options
Door Style:
2-Split Inner Door
Single Glass Inner Door
Oxygen Control:
1 to 19% O2 Control
None
Unit Size:
Each
Description
- 48 Liter (1.7 ft3) provides mid-sized capacity within a small footprint
- Options: High Temp. Disinfection (4 hour at 120 °C), O2 control (e.g. for hypoxic studies), 2-split inner glass door
- Fanless design offers more usable space
- Six-sided direct heating system provides a uniform incubated environment
- Deep-drawn, easy-to-clean stainless steel chamber without seams or welds
- Low gas consumption
- Integrated alarm relay
Specifications
Specifications
| Description | UK/HKG Power Supply |
| CO2 Sensor Technology | IR Sensor |
| Oxygen Control | None |
| Door Style | Single Glass Inner Door |