Learn More
Poulten & Graf GmbH FORTUNA™ Burette Bang pattern
Achieve better accuracy and repeatability, especially in titrations. Poulten & Graf GmbH FORTUNA™ Burette Bang pattern delivers accurate and precise volumes.
Brand:  Poulten & Graf GmbH 1 03034G
 View more versions of this product
Product Code. 10771693
  • € 134.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comformity approved
  • Blue graduation
  • With glass key in upper stopcock
  • With straight glass stopcock
Specifications
Specifications
10 mL
0.02 mL
Blue
Glass
Class AS
Borosilicate Glass
Burette Bang Pattern
Straight
±0.02 mL
EN ISO 385
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers