Learn More
Poulten & Graf GmbH FORTUNA™ Burette Bang pattern
Achieve better accuracy and repeatability, especially in titrations. Poulten & Graf GmbH FORTUNA™ Burette Bang pattern delivers accurate and precise volumes.
Brand: Poulten & Graf GmbH 1 03023G
This item is not returnable.
View return policy
Description
- Comformity approved
- Blue graduation
- With glass key in upper stopcock
- With straight glass stopcock
Specifications
2 mL | |
0.01 mL | |
0.01 mL |
Borosilicate Glass | |
Burette Bang pattern | |
EN ISO 385 |