Learn More
Schneider Gerd™ Forceps
Brand: Schneider Gerd™ 127
This item is not returnable.
View return policy
Specifications
Bent | |
300 mm | |
Forceps | |
For Laboratory use |
Product Content Correction
Product Title
Schneider Gerd™ Forceps > Length: 300mm