Learn More
Grant Instruments™ Flask Platforms for use with ES-20/80
The Grant™ interchangeable flask platforms are for use with the ES-20/80 Orbital Shaker-Incubator. The platforms have clamps to securely hold the flasks in position during vigorous mixing.
Brand:  Grant Instruments™ HSP-30/100
 View more versions of this product
Product Code. 17755019
  • € 902.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Interchangeable
  • Clamps securely hold flasks in place during vigorous mixing
  • Optional accessories for the ES-20/80 Orbital Shaker-Incubator
TRUSTED_SUSTAINABILITY
Specifications
Specifications
30 x 100 to 150 mL Flasks
7.3 lb.
Platform for 100 mL Flasks (30 x 100-150 mL Flasks. ES-20/80 and ES-20/80 C)
ES-20/80 and ES-20/80C
36 x 40 x 10 cm
3.3 kg
HSP-30/100
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Flask Platforms for use with ES-20/80 > Holds: 30 x 100 to 150 mL Flasks