Learn More
Cytiva Support Screens for XK and HiScale Columns
For use with XK and HiScale Columns
Brand:  Cytiva 19-0651-01
39.90 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10715152
  • € 42.00 / Pack of 5
In Stock
to see stock.
Explore more special offers
Description
  • Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Specifications
16 mm
Polypropylene
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Support Screens for XK and HiScale Columns > I.D.: 16mm