Learn More
Liebherr™ FKV503 Model Stainless Steel/Glass Door Refrigerator
Brand:  Liebherr™ FKV 503
Product Code. 15106192
Specifications
General Purpose Refrigerator
2°C to 12°C
HardLine
3
450 mm
425 mm
220 - 240 V 50 Hz
0.763 kWh/Annual
45 L
R 600a
3
Mechanical
612 mm
220/240 V
50 Hz
31 kg
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Liebherr™ FKV503 Model Stainless Steel/Glass Door Refrigerator > Capacity: 45L