Learn More
Grant Instruments™ External Probe
External probes for monitoring and controlling temperature of remote loads.
€ 244.00 - € 251.00
Specifications
Length (English) 3 m
View More Specs
Products 2
Product Code Brand Material Price Quantity & Availability  
Product Code Brand Material Price Quantity & Availability  
15449224
View Documents
Grant Instruments™
SEP
Stainless Steel
€ 244.00
Each
In Stock
to see stock.
Add to basket
15429224
View Documents
Grant Instruments™
PEP
Plastic
€ 251.00
Each
In Stock
to see stock.
Add to basket
Description
Description
Grant Instruments temperature probe (Pt1000) external for use with LT ecocool 150
  • 3 m cable
Specifications
Specifications
3 m
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Grant Instruments™ External Probe