Learn More
Grant Instruments™ External Probe
Click to view available options
Material:
Plastic
Stainless Steel
Unit Size:
Each
Description
Grant Instruments temperature probe (Pt1000) external for use with LT ecocool 150
- 3 m cable
Specifications
Specifications
| Material | Plastic |
| Length (English) | 3 m |