Learn More
Tuttnauer Extend Cycle Time
Special program to work with extended sterilization times of more than 99 minutes. Must specify sterilization temperature (121°C or 134°C) and length of sterilization time.
Brand:  Tuttnauer ADD222-0458
Product Code. 17819538
  • € 1685.00 / Each
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Description
  • Special program to work with extended sterilization times of more than 99 minutes.
  • Must specify sterilization temperature (121°C or 134°C) and length of sterilization time.
Specifications
Specifications
Software
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Extend Cycle Time > Extend Cycle Time