Learn More
Tuttnauer Air Compressor
The Air Compressor introduces pressurized air into the chamber for rapid cooling of liquids for models with a fast cooling system. Not needed if air pressure source is available on site.
Brand:  Tuttnauer PUM059-0023
 View more versions of this product
Product Code. 17869538
  • € 1715.00 / Each
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Description
  • Introduces pressurized air into the chamber for rapid cooling of liquids for models with a fast cooling system.
  • Not needed if air pressure source is available on site.
Specifications
Specifications
Air Compressor
50 Hz
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Air Compressor > Air Compressor; Frequency: 50 Hz