missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 3,5-bis(trifluoromethyl)phenylacetate, 98%, Thermo Scientific™ Product Code.: 11485530

Ethyl 3,5-bis(trifluoromethyl)phenylacetate, 98%, Thermo Scientific™

Product Code. 11485530
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11485530

Brand: Thermo Scientific Alfa Aesar H25820.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144632-97-3
Molecular Formula C12H10F6O2
Molecular Weight (g/mol) 300.2
MDL Number MFCD08689894
InChI Key ICTVPOAERFKXSS-UHFFFAOYSA-N
Synonym ethyl 3,5-bis trifluoromethyl phenyl acetate, ethyl 3,5-bis trifluoromethyl phenylacetate, 3,5-bis trifluoromethyl phenylacetic acid ethyl ester, ethyl 2-3,5-bis trifluoromethyl phenyl acetate, ethyl 2-3,5-bis trifluoromethyl phenyl ethanoate, 3,5-di trifluoromethyl phenylacetic acid ethyl ester, 2-3,5-bis trifluoromethyl phenyl acetic acid ethyl esterShow MoreShow Less
PubChem CID 42552891
IUPAC Name ethyl 2-[3,5-bis(trifluoromethyl)phenyl]acetate
SMILES CCOC(=O)CC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Refractive Index 1.417
Quantity 1 g
Formula Weight 300.2
Percent Purity 98%
Chemical Name or Material Ethyl 3,5-bis(trifluoromethyl)phenylacetate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10182370 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10147815 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberE30Y012 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 3,5-bis(trifluoromethyl)phenylacetate, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.