Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With Ground Joint, Amber
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
€ 366.76 - € 1224.30
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11913381
|
Lenz Laborglasintrumente
03023058 |
500 mL |
€ 528.94
Pack of 10 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
11923381
|
Lenz Laborglasintrumente
03023070 |
1 L |
€ 692.18
Pack of 10 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
11933381
|
Lenz Laborglasintrumente
03023073 |
2 L |
€ 1224.30
Pack of 10 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
11993371
|
Lenz Laborglasintrumente
03023046 |
200 mL |
€ 366.76
Pack of 10 |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN EN ISO 4797
- With scale (approx. volume /ml)
- With inscription label
- Material: DURAN™
Specifications
NS 29/32 |