Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With Ground Joint, Amber
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03023058
 View more versions of this product
Product Code. 11913381
  • € 559.00 / Pack of 10
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
500 mL
NS 29/32
105 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers