Learn More
Equilin, MedChemExpress
MedChemExpress Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.
Brand: Medchem Express HY-B1176
Description
Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.Specifications
Equilin | |
White | |
C18H20O2 | |
7-Dehydroestrone | |
C[C@]1([C@](CC2)([H])C3=CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O | |
268.35 | |
Research |
474-86-2 | |
Research | |
50 mg | |
DMSO : 100 mg/mL (372.65 mM; Need ultrasonic) | |
268.35 | |
99.0% | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only