Learn More
Eprinomectin, MedChemExpress
MedChemExpress Eprinomectin(MK-397) is an avermectin selected for development as a topical endectocide; has anthelmintic, insecticidal and miticidal activity.
POA - € 151.50
Specifications
Chemical Name or Material | Eprinomectin |
---|---|
CAS | 123997-26-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C50H75NO14 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16475936
|
Medchem Express
HY-12643 |
100 mg |
€ 151.50
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16431646
|
Medchem Express
HY-12643 |
500 mg |
POA
500mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Eprinomectin(MK-397) is an avermectin selected for development as a topical endectocideSpecifications
Eprinomectin | |
White | |
C50H75NO14 | |
DMSO : ≥ 100 mg/mL (109.39 mM) | |
914.13 | |
98.0% | |
Solid |
123997-26-2 | |
Research | |
MK-397 | |
C[C@H]1[C@@H](NC(C)=O)[C@@H](OC)C[C@H](O[C@@H]2[C@H](C)O[C@@H](O[C@@H](/C(C)=C/C[C@@H](C3)OC4(O[C@@H]([C@@H](CC)C)[C@H](C)C=C4)C[C@H]3OC5=O)[C@@H](/C=C/C=C6CO[C@@H]7[C@@]\6(O)[C@H]5C=C(C)[C@H]7O)C)C[C@@H]2OC)O1 | |
914.13 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H225∣H302+H312+H332∣H319
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.