Learn More
Eprinomectin, MedChemExpress
MedChemExpress Eprinomectin(MK-397) is an avermectin selected for development as a topical endectocide; has anthelmintic, insecticidal and miticidal activity.
Brand: Medchem Express HY-12643
This item is not returnable.
View return policy
Description
Eprinomectin(MK-397) is an avermectin selected for development as a topical endectocideSpecifications
Eprinomectin | |
White | |
C50H75NO14 | |
MK-397 | |
C[C@H]1[C@@H](NC(C)=O)[C@@H](OC)C[C@H](O[C@@H]2[C@H](C)O[C@@H](O[C@@H](/C(C)=C/C[C@@H](C3)OC4(O[C@@H]([C@@H](CC)C)[C@H](C)C=C4)C[C@H]3OC5=O)[C@@H](/C=C/C=C6CO[C@@H]7[C@@]\6(O)[C@H]5C=C(C)[C@H]7O)C)C[C@@H]2OC)O1 | |
914.13 | |
Research | |
COVID-19-immunoregulation |
123997-26-2 | |
Research | |
500 mg | |
DMSO : ≥ 100 mg/mL (109.39 mM) | |
914.13 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only