Learn More
Schneider Gerd™ Double Spatula
Use this spatula for a variety of pharmaceutical applications. Schneider Gerd™ Double Spatulas are ideal for use with a wide range of compounds.
Brand:  Schneider Gerd™ 8301
 View more versions of this product
Product Code. 10001001
  • € 2.00 / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Description
  • Spatulas are ideal for working with powders, stirring and mixing
  • Two end shapes offer increased versatility
  • Perfect for miniscule amounts of chemicals
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
1 Pk.
8 mm
130 mm
Double Ended Spatula
130 x 8 mm
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Schneider Gerd™ Double Spatula > Length: 130mm