Learn More
Schneider Gerd™ Double Spatula
Product Code. p-8008625
Click to view available options
Length (Metric):
130 mm
185 mm
250 mm
300 mm
Unit Size:
Each
Product Code. 10051031
Brand: Schneider Gerd™ 8305
Only null left
Add to basket
Add to basket
Use this spatula for a variety of pharmaceutical applications. Schneider Gerd™ Double Spatulas are ideal for use with a wide range of compounds.
  • Spatulas are ideal for working with powders, stirring and mixing
  • Two end shapes offer increased versatility
  • Perfect for miniscule amounts of chemicals
  • Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Quantity 1 Pk.
Product Type Double Ended Spatula
Width (Metric) Blade 11 mm
Dimensions (L x W) 250 x 11 mm
Length (Metric) 250 mm