Learn More
Disposable Luer-Slip Two-Part Syringe
Safe application of medication is ensured by the high quality of these needles. Henke Sass Wolf Hsw™ Disposable Luer-Slip Two-Part Syringes are designed for optimized user handling to ensure patient safety.
€ 4.96 - € 8.96
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | DIN, EN, ISO and FDA standards, Quality management system certified according to ISO 13485:2003 / AC 2007, CE-marked |
Material | Polypropylene |
Color | Transparent Barrel |
Description | Smudge-proof scale |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10018740
|
Henke Sass Wolf Hsw
HSW4020000DO |
2 to 3 mL |
€ 4.96
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10212521
|
Henke Sass Wolf Hsw
HSW4050000DO |
5 to 6 mL |
€ 6.98
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10549111
|
Henke Sass Wolf Hsw
HSW4100000V0 |
10 to 12 mL | |||||||
Description
- Available from sizes 1 to 50mL (60mL)
- Smooth flow, tight, high transparency barrel
- No rubber, styrene or DEHP
- Latex- and silicone-oil-free
- Pyrogen-free, PVC-free, non-toxic
- Sterile, individually blister strip packed
- Your first choice for an inert syringe
- Defined position of the plunger at volume “0” to feel when plunger is completely inserted
- LDS: no dead space for 1mL syringe
- Extended graduation for wider range of application
- Positive safety stop to prevent accidental spills
- 1 to 20mL (24mL) available also with by-packed needle
- 2 to 20mL available as Oral syringe with purple plunger
- 2mL to 50 mL (60mL) also available with Luer Lock
- 30mL and 50mL also available with Catheter tip
- All sizes available in non-sterile version (bulk) upon request
Specifications
Autoclavable | |
Polypropylene | |
Smudge-proof scale | |
Luer-slip 2-part Syringe | |
100/Pk. | |
Yes |
DIN, EN, ISO and FDA standards, Quality management system certified according to ISO 13485:2003 / AC 2007, CE-marked | |
Transparent Barrel | |
Yes | |
Single-use | |
Leur Slip | |
Sterile |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.