Learn More
Fisherbrand™ Catalyst PharMed BPT Formulation Pump Tubing
Available in standard 15m Lengths.
Brand:  Fisherbrand™ 97000-23
 View more versions of this product
Product Code. 15511305
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
    • Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
    • Available in both thin and thick wall configurations
Specifications
Specifications
4.8 mm
1.6 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers