missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DICER1 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
Antigen | DICER1 |
---|---|
Clone | OTI2B1 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17540003
|
Origene Technologies
TA801636S |
30 μL |
€ 242.00
30µL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17520003
|
Origene Technologies
TA801636 |
100 μL |
€ 540.87
100µL |
Estimated Shipment: 15-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DICER1 Monoclonal antibody specifically detects DICER1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DICER1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Q9UPY3 | |
IgG2a | |
-20° C, Avoid Freeze/Thaw Cycles |
OTI2B1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
23405 | |
Primary | |
DICER1 |