missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DICER1 Monoclonal Antibody (OTI2B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801636S
Additional Details : Weight : 0.05000kg
Description
DICER1 Monoclonal antibody specifically detects DICER1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
DICER1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9UPY3 | |
Mouse | |
Affinity chromatography | |
RUO | |
23405 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Immunohistochemistry (Paraffin) | |
OTI2B1 | |
Unconjugated | |
DICER1 | |
1110006F08Rik; D12Ertd7e; DCR; DCR1; DCR-1; Dicer; dicer 1 ribonuclease III; dicer 1, double-stranded RNA-specific endoribonuclease; dicer 1, ribonuclease III; dicer 1, ribonuclease type III; Dicer1; Dicer1, Dcr-1 homolog; Dicer1e; double-strand-specific ribonuclease mDCR-1; endoribonuclease Dicer; Helicase MOI; helicase with RNAse motif; helicase-moi; HERNA; K12H4.8-LIKE; KIAA0928; mKIAA0928; MNG1; RMSE2 | |
Human recombinant protein fragment corresponding to amino acids 1666-1922 of human DICER1 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |