Learn More
Desloratadine, MedChemExpress
MedChemExpress Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.
€ 90.45 - € 495.45
Specifications
Chemical Name or Material | Desloratadine |
---|---|
Physical Form | Solid |
CAS | 100643-71-8 |
Color | Off-White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498356
|
Medchem Express
HY-B0539 |
100 mg |
€ 138.00
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16482166
|
Medchem Express
HY-B0539 |
500 mg |
€ 495.45
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16404397
|
Medchem Express
HY-B0539 |
50 mg |
€ 90.45
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.Specifications
Desloratadine | |
100643-71-8 | |
Research | |
COVID-19-immunoregulation | |
DMSO : 25 mg/mL (80.43 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.16 mM; Need ultrasonic) | |
310.82 | |
99.98% |
Solid | |
Off-White | |
C19H19ClN2 | |
Sch34117 | |
ClC1=CC=C2C(CCC3=CC=CN=C3/C2=C4CCNCC/4)=C1 | |
310.82 | |
Research |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.