Learn More
Desloratadine, MedChemExpress
MedChemExpress Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.
Brand: Medchem Express HY-B0539
This item is not returnable.
View return policy
Description
Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.Specifications
Desloratadine | |
100643-71-8 | |
500 mg | |
C19H19ClN2 | |
Sch34117 | |
ClC1=CC=C2C(CCC3=CC=CN=C3/C2=C4CCNCC/4)=C1 | |
310.82 | |
Research |
Solid | |
Off-White | |
Research | |
COVID-19-immunoregulation | |
DMSO : 25 mg/mL (80.43 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.16 mM; Need ultrasonic) | |
310.82 | |
99.98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only