Learn More
Desloratadine, MedChemExpress
MedChemExpress Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.
Brand:  Medchem Express HY-B0539
Encompass_Preferred
 View more versions of this product
Product Code. 16482166
  • € 495.45 / 500mg
Explore more special offers
Description
Description
Desloratadine (Sch34117) is the orally active major metabolite of the nonsedating H1-antihistamine Loratadine. Desloratadine is a selective H1-receptor antagonist that has anti-allergic and anti-inflammatory activities.
Specifications
Specifications
Desloratadine
100643-71-8
500 mg
C19H19ClN2
Sch34117
ClC1=CC=C2C(CCC3=CC=CN=C3/C2=C4CCNCC/4)=C1
310.82
Research
Solid
Off-White
Research
COVID-19-immunoregulation
DMSO : 25 mg/mL (80.43 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.16 mM; Need ultrasonic)
310.82
99.98%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only