Learn More
Davercin, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
Description
Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
Specifications
Specifications
| Chemical Name or Material | Davercin |
| CAS | 55224-05-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C38H65NO14 |
| Quantity | 10 mg |
| Synonym | Erythromycin Cyclocarbonate |
| Solubility Information | DMSO : ≥ 50 mg/mL (65.80 mM) |
| SMILES | C[C@]([C@H](OC([C@@H]([C@H]([C@H](C)[C@H]1O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)C)=O)CC)(O4)[C@]([C@H](C([C@@H](C[C@]1(O)C)C)=O)C)([H])OC4=O |
| Molecular Weight (g/mol) | 759.92 |
| Show More |
Research purposes only