missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Custom Silencer™ siRNA

Product Code. 15521521
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
Each
This item is not returnable. View return policy

Product Code. 15521521

Brand: Ambion™ AM16204

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Custom Silencer™ siRNA

RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room temperature
Description Custom siRNA, 40nmol, HPLC Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Product Type Custom siRNA
Purity HPLC
Quantity 40 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.