Learn More
Ambion™ Custom Silencer™ siRNA
Product Code. 15511521
Click to view available options
Quantity:
1 umol
100 nmol
20 nmol
40 nmol
Unit Size:
100nmol
1µmol
20nmol
40nmol
Each
Product Code. 15511521
Brand: Ambion™ AM16106
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Custom Silencer™ siRNA
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Custom siRNA, 40nmol, Liquid, Standard Purity, Water Soluble, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA
Product Type Custom siRNA
Purity Standard
Quantity 40 nmol
Form Liquid