Learn More
Ambion™ Custom Select siRNA
Product Code. 15561064
Click to view available options
Quantity:
1 μmol
10 μmol
20 nmol
250 nmol
40 nmol
5 nmol
Conditionnement:
20nmol
250nmol
40nmol
5nmol
Each
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 15561064
Marque: Ambion™ 4404013
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Custom Select siRNA
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
TRUSTED_SUSTAINABILITY
Spécification
Content And Storage Room temperature
Description Custom Select siRNA, 250nmol, HPLC Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities
For Use With (Application) Knocking down gene expression in order to study protein function in a wide range of cell types
Product Type Custom Select siRNA
Purity HPLC
Quantity 250 nmol