Learn More
CUDC-101, MedChemExpress
MedChemExpress CUDC-101 is a potent inhibitor of HDAC, EGFR, and HER2 with IC50s of 4.4, 2.4, and 15.7 nM, respectively.
Brand:  Medchem Express HY-10223
Encompass_Preferred
 View more versions of this product
Product Code. 16414796
  • € 742.50 / 100mg
Explore more special offers
Description
Description
CUDC-101 is a potent inhibitor of HDAC, EGFR, and HER2 with IC50s of 4.4, 2.4, and 15.7 nM, respectively.
Specifications
Specifications
CUDC-101
White
C24H26N4O4
DMSO : 25 mg/mL (57.54 mM; Need ultrasonic)
434.49
97.0%
Solid
1012054-59-9
Research
100 mg
C#CC1=CC=CC(NC2=NC=NC3=CC(OC)=C(C=C23)OCCCCCCC(NO)=O)=C1
434.49
Research
Cancer-Kinase/protease
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
CUDC-101, MedChemExpress
Research purposes only