Learn More
Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
Brand: Corning™ 9998-241
Description
Made from fabricated specially formulated resin to resist the effects of temperature and steam in autoclaving. Corning™ Black Phenolic Threaded Screw Caps with PTFE Liners provide a highly inert sealing face.
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
Specifications
24-410 | |
Screw Cap | |
Phenolic | |
18.5 mm | |
144/Cs. |
PTFE | |
Autoclavable | |
Black | |
144/Pk. | |
PTFE, Rubber |
Product Suggestions
Customers who viewed this item also viewed