Learn More
Corning™ 384-Well, Non-Treated, Flat-Bottom Microplate

Color:Black; Sterility:Non-sterile; Lid:Without Lid
Brand: Corning™ 3821BC
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.

Specifications
Standard | |
0.05 cm2 | |
Black | |
384 | |
Non-sterile | |
50 μL | |
With generic barcode | |
10/Pk. | |
Clear |
Without Lid | |
16 x 24 | |
Polystyrene | |
5 to 40 μL | |
Non-treated | |
Flat Bottom (F-well) | |
50/Cs. | |
50/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.