Learn More
Levetiracetam, MedChemExpress
MedChemExpress Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.
Brand:  Medchem Express HY-B0106
Encompass
 View more versions of this product
Product Code. 16434237
  • POA / 50mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Levetiracetam, an antiepileptic agent, binds the synaptic vesicle protein SV2A. Levetiracetam enhances Temozolomide effect on glioblastoma stem cell proliferation and apoptosis. Levetiracetam modulates HDAC levels ultimately silencing MGMT, thus increasing Temozolomide effectiveness. A chemosensitizer agent.
Specifications
Specifications
Levetiracetam
White
50 mg
H2O : ≥ 85 mg/mL (499.38 mM) ∣DMSO : ≥ 85 mg/mL (499.38 mM)
170.21
Research
102767-28-2
Research
UCB L059
NC([C@@H](N(CCC1)C1=O)CC)=O
99.21%
Powder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Levetiracetam, MedChemExpress
Research purposes only