Learn More
Hach Lange™ COD Cuvette Test
€ 145.00 - € 148.00
Product Code | Brand | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Price | Quantity & Availability | |||||
10530323
|
Hach Lange™
LCI400 |
0 to 1000mg/L O2 |
€ 148.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10671000
|
Hach Lange™
LCK114 |
150 to 1000 mg/L O2 |
€ 145.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Dichromate | |
COD Cuvette Test |
Safety and Handling
missing translation for 'shelfLife' : 30 Months
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- CSB/COD/DCO
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Germ cell mutagenicity Category 1B
- Carcinogenicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H290-May be corrosive to metals.
- H311-Toxic in contact with skin.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH208-Contains potassium dichromate. May produce an allergic reaction.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid ... %, Mercury(II) sulfate, Silver sulfate, potassium dichromate
Product Title
Hach Lange™ COD Cuvette Test