Learn More
Hach Lange™ COD Cuvette Test
Brand:  Hach Lange™ LCK114
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10671000
  • € 145.00 / Each
Explore more special offers
Specifications
ISO 6060-1989, DIN 38409-H41-H44
25
Dichromate
For Automatic Recognition in the Photometer
150 to 1000 mg/L O2
COD Cuvette Test
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hach Lange™ COD Cuvette Test > Range: 150 to 1000 mg/L O2