Learn More
Hirschmann™ Class A Amber Glass White Graduated Measuring Flasks
Provided in amber glass for the protection of light-sensitive contents.
Brand:  Hirschmann™ 264018027
 View more versions of this product
Product Code. 10727803
  • € 80.40 / Pack of 2
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • White graduation
  • Class A
  • With dated batch identification
  • Standard ground and PE stopper
  • Manufactured from DURAN™ borosilicate glass for high chemical and thermal resistance
  • USP certified
Specifications
Specifications
Polyethylene
100 mL
Liquid handling of light-sensitive products
Ground 12/21
A
Stopper
Amber
Borosilicate Glass
170 ± 5 mm
Yes
Flat
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers