Learn More
Hirschmann™ Class A Amber Glass White Graduated Measuring Flasks
Provided in amber glass for the protection of light-sensitive contents.
Brand:  Hirschmann™ 264018927
 View more versions of this product
Product Code. 10597383
  • € 138.00 / Pack of 2
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Description
  • White graduation
  • Class A
  • With dated batch identification
  • Standard ground and PE stopper
  • Manufactured from DURAN™ borosilicate glass for high chemical and thermal resistance
  • USP certified
Specifications
Specifications
Polyethylene
500 mL
Liquid handling of light-sensitive products
Ground 19/26
A
Stopper
Amber
Borosilicate Glass
260 ± 5 mm
Yes
Flat