Learn More
Tocris Bioscience™ CIA 1
COUP-TFII inhibitor
€ 188.00 - € 715.00
Specifications
Chemical Name or Material N-[2-(3,4-Dimethoxyphenyl)ethyl]-6-methylthieno[2,3-d]pyrimidin-4-amine hydrochloride
CAS 1049691-47-5
Target Additional Nuclear Receptor Compounds
Molecular Formula C17H19N3O2S.HCl
Purity 0.98
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18727486
View Documents
Tocris Bioscience™
7494/10
10 mg
€ 188.00
10mg
18779146
View Documents
Tocris Bioscience™
7494/50
50 mg
€ 715.00
50mg
Description
Description
CIA 1 is an inhibitor of the nuclear receptor COUP-TFII (IC50 ranged from 1.2 μM to 7.6 μM, in prostate cancer cell lines). In vitro, CIA 1 inhibits growth of multiple prostate cancer cell lines and suppresses COUP-TFII activity to regulate its target genes. In vivo, CIA 1 reduces tumor growth in prostate cancer xenograft mouse models.
Specifications
Specifications
N-[2-(3,4-Dimethoxyphenyl)ethyl]-6-methylthieno[2,3-d]pyrimidin-4-amine hydrochloride
Additional Nuclear Receptor Compounds
0.98
1049691-47-5
C17H19N3O2S.HCl
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Tocris Bioscience™ CIA 1