Learn More
Macherey-Nagel™ Chromafix™ C18 ec Cartridge
Shop All Macherey-Nagel Products
Click to view available options
Bed Weight:
270 mg
530 mg
950 mg
missing translation for 'unitSize'
Pack of 50
Specifications
Specifications
| Base Material | Silica |
| Carbon Load | 14% |
| Bed Weight | 530 mg |
| For Use With (Application) | Non-Polar Compounds, Aflatoxins, Amphetamines, Antibiotics, Antiepileptics, Barbiturates, Caffeine, Drugs, Preservatives, Fatty Acids, Nicotine, PAHs, Pesticides, PCBs, Heavy Metals, Vitamins, Very Well Suited for Desalting of Samples |
| Particle Size | 45 μm |
| Sorbent | C18 ec |
| Type | Chromafix C18 ec Cartridge |
| Endcapped | Yes |
| Pore Size | 60 Å |
| Quantity | 50 Pack |
| Show More |