Learn More
Macherey-Nagel™ CHROMABOND SPE Vacuum Manifold
The complete CHROMABOND SPE vacuum manifold is the perfect choice for speeding up sample processing of up to 12 positions simultaneously.
Brand:  Macherey-Nagel™ 730150N
839.32 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16099311
  • € 990.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Complete vacuum manifold system
Specifications
Specifications
Vacuum Manifold, Complete System
Sample Preparation
1x Glass cabinet, 1x lid (complete, incl. 4x feet, Luer fittings, 1x lid gasket), 12x exchangeable needles (PP), 1x vacuum gauge, 1x control valve, 12x Luer valves (stopcocks), 12x caps, 1x variable rack (6 plates), 1x PP tank
12 Positions
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ CHROMABOND SPE Vacuum Manifold > SPE Vacuum Manifold; Number of Ports: 12 Positions