Learn More
Azpack™ Matte Vellum Paper Labels
Adhere to a variety of surfaces. Azpack™ Matte Vellum Paper Labels can be used with a thermal transfer printer to create labels.
Brand:  Azpack™ 10193474
 View more versions of this product
Product Code. 10193474
  • € 30.10 / Pack of 1000
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Matte white vellum paper
  • Thermal transfer
  • Glassine finish
  • Permanent adhesive backing
Specifications
Specifications
Deep die cut wide edge
White
Vellum
24 x 13 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Azpack™ Matte Vellum Paper Labels > White; Dimensions: 24L x 13mmW