Learn More
Azpack™ Matte Vellum Paper Labels
Adhere to a variety of surfaces. Azpack™ Matte Vellum Paper Labels can be used with a thermal transfer printer to create labels.
Brand: Azpack™ 10491274
This item is not returnable.
View return policy
Description
- Matte white vellum paper
- Thermal transfer
- Glassine finish
- Permanent adhesive backing
Specifications
Deep die cut wide edge | |
Vellum | |
49 x 25.4 mm |
White | |
500/Pk. |
Product Content Correction
Product Title
Azpack™ Matte Vellum Paper Labels > Dimensions: 49L x 25.4mmW