Learn More
Chroma Gesellschaft™ 8mm Plier Style Decapper
Brand:  JG Finneran Associates 9310-08
Product Code. 10537932
  • € 147.00 / Each
Estimated Shipment: 25-07-2025
to see stock.
Explore more special offers
Specifications
8 mm
Plier-style Decapper
8 mm Caps
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Chroma Gesellschaft™ 8mm Plier Style Decapper >