Learn More
Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials
Developed for use in any turbidimeter. Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials are true Formazin dilutions in individual sealed vials of <0.1, 20, 200, 1000, 4000 NTU and 7500 NTU (2662105 only) levels.
Brand:  Hach™ 2659505
 View more versions of this product
Product Code. 10346162
  • € 738.00 / 1 set
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Developed for use in any turbidimeter. Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials are true Formazin dilutions in individual sealed vials of <0.1, 20, 200, 1000, 4000 NTU and 7500 NTU (2662105 only) levels.
  • Precise, Pre-mixed Formazin Solutions: Provides accurate turbidimeter calibrations
  • Based on Formazin Primary Standards: Contains the same light scattering polymer and performs equivalently
  • Enhanced Stability Saves Time and Labor: Repeatable results without preparing standards in the lab
  • Confidence in Results: Based on decades of experience in the drinking water industry
  • Any Turbidity Meter: Can be used with 2100N/2100AN or 2100N/2100AN IS Turbidimeter
Specifications
Specifications
Stablcal Turbidity Standards Calibration Kit
Premixed, sealed vials
<0.1, 20, 200, 1000, 4000, 7500 NTU
2100AN/AN IS
Calibration of turbidimeters
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hach™ Stablcal™ Turbidity Standards Calibration Kit, Sealed Vials > 0-7500 NTU; For use with 2100AN/AN IS Turbidimeters