Learn More
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X)
Cocktail of phorbol 12-myristate 13-acetate (PMA) and ionomycin. Treatment with PMA and ionomycin is sufficient to induce activation of many cell types to produce cytokines.
Brand:  Invitrogen™ 00-4970-93
70.35 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.
 View more versions of this product
Product Code. 15452024
  • € 105.00 / 100µL
Explore more special offers
Description
Description
  • Can be used to induce cytokine production in in vitro cell culture for subsequent detection in immunoassays
  • Can also be used in conjunction with the Protein Transport Inhibitor Cocktail (500X) (cat. 00-4980) for intracellular retention of the induced cytokine expression
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 μL
100μL
eBioscience™
Cell Stimulation Cocktail
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2916128 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number00-4970-93
Lot Number2430455 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number00-4970-93
Lot Number2504666 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number00-4970-93
Lot Number2504665 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number00-4970-93
Lot Number2546957 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number00-4970-93
Special Offers
Special Offers
Product Content Correction
Product Title
Invitrogen™ eBioscience™ Cell Stimulation Cocktail (500X) > 100uL