Learn More
SPEX™ Cap-Mat for Round-Well Plate
SPEX™ Cap-Mat is a sealing mat specifically designed for the 2211 96-Well Titer Plate.
Brand:  SPEX™ 2211
Product Code. 13561350
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Tightly seals titer plate wells
  • Prevents spills and well-to-well contamination
  • Made of rugged silicone rubber
  • May be sterilized and reused
Specifications
Specifications
2211 96-Well Titer Plate
Silicone Rubber
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SPEX™ Cap-Mat for Round-Well Plate > Material: Silicone Rubber