missing translation for 'onlineSavingsMsg'
Learn More

BV6, MedChemExpress

MedChemExpress BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.

€ 196.50 - € 1630.80

Specifications

Chemical Name or Material BV6
CAS 1001600-56-1
Color White
Purity Grade Notes Research
Molecular Formula C70H96N10O8
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16483167
Encompass_Preferred
View Documents
Medchem Express
HY-16701
50 mg
€ 1063.80
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16449677
Encompass
View Documents
Medchem Express
HY-16701
5 mg
€ 196.50
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16447146
Encompass_Preferred
View Documents
Medchem Express
HY-16701
100 mg
€ 1630.80
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16407148
Encompass_Preferred
View Documents
Medchem Express
HY-16701
10 mg
€ 306.45
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.
Specifications

Specifications

BV6
White
C70H96N10O8
O=C(NCCCCCCNC([C@@H](NC([C@H](CCC1)N1C([C@H](C2CCCCC2)NC([C@H](C)NC)=O)=O)=O)C(C3=CC=CC=C3)C4=CC=CC=C4)=O)[C@@H](NC([C@H](CCC5)N5C([C@H](C6CCCCC6)NC([C@H](C)NC)=O)=O)=O)C(C7=CC=CC=C7)C8=CC=CC=C8
1205.57
Research
Cancer-programmed cell death
1001600-56-1
Research
DMSO : ≥ 58 mg/mL (48.11 mM)
1205.57
99.84%
Solid
Videos
Safety and Handling

Safety and Handling

H302∣H315∣H319∣H335

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.