Learn More
BV6, MedChemExpress
MedChemExpress BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.
Brand: Medchem Express HY-16701
This item is not returnable.
View return policy
Description
BV6 is an antagonist of cIAP1 and XIAP, members of the inhibitors of apoptosis (IAP) family.Specifications
BV6 | |
White | |
C70H96N10O8 | |
DMSO : ≥ 58 mg/mL (48.11 mM) | |
1205.57 | |
99.84% | |
Solid |
1001600-56-1 | |
Research | |
10 mg | |
O=C(NCCCCCCNC([C@@H](NC([C@H](CCC1)N1C([C@H](C2CCCCC2)NC([C@H](C)NC)=O)=O)=O)C(C3=CC=CC=C3)C4=CC=CC=C4)=O)[C@@H](NC([C@H](CCC5)N5C([C@H](C6CCCCC6)NC([C@H](C)NC)=O)=O)=O)C(C7=CC=CC=C7)C8=CC=CC=C8 | |
1205.57 | |
Research | |
Cancer-programmed cell death |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
BV6, MedChemExpress
Research purposes only