Learn More
Buerkle™ Polyethylene Spigot Jumbo with Tap
Spigot with tap for R2in.barrel thread, 50 mm flow, for viscous media such as lacquers, glue, shampoo.
Brand: Buerkle™ 0534-0020
Description
- Cost-effective tap
- PE, brown
- Seal made of EPT
Specifications
R 2 in. barrel thread | |
Brown | |
PE, EPT | |
Spigot |